Learn More
Bellco Glass, Inc. Stainless Steel Bioreactor Headplate, 3L
SDP
Catalog No. 501944897
Encompass_Preferred
3L bioreactor headplate . Manufactured from 316l stainless steel with ra 32 finish or better inside and out. Fittings are sold separately as part of the system. MADE IN THE USA.
Catalog No. 50-194-4897 Supplier Bellco Glass, Inc. Supplier No. 196488503
3L bioreactor headplate . Manufactured from 316l stainless steel with ra 32 finish or better inside and out. Fittings are sold separately as part of the system. MADE IN THE USA.