Learn More
Bellco Glass, Inc. Stainless Steel Bioreactor Headplate,7L
SDP
Catalog No. 501944899
Encompass_Preferred
7L bioreactor headplate. Manufactured from 316l stainless steel with ra 32 finish or better inside and out. Fittings are sold separately as part of the system. MADE IN THE USA.
Catalog No. 50-194-4899 Supplier Bellco Glass, Inc. Supplier No. 196488507
7L bioreactor headplate. Manufactured from 316l stainless steel with ra 32 finish or better inside and out. Fittings are sold separately as part of the system. MADE IN THE USA.