Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Caresfield SST Blood Collection Tube, 13 x 100 mm, Gold Top
SDP

Catalog No. 22246102
Change view
Click to view available options
Capacity (Metric):
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
22-246-102 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-246-102 Supplier Caresfield Supplier No. 63261
Add to Cart
Add to Cart

Gold-top SST blood collection tube designed for serum specimen collection.

  • 5 mL collection capacity
  • Tube dimensions: 13 x 100 mm
  • Gold-top design for easy identification

Specifications

Color Gold
Product Type Blood Collection Tube
Additive Clot Activator and Gel
Material Plastic
Sterility Sterile
Closure Type Cap
Capacity (Metric) 5 mL
Diameter (Metric) 13 mm
Quantity 1000/Cs.
Length (Metric) 100 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.