Learn More
Bellco Glass, Inc. Stainless Steel Closure,13mm (Lanced)
SDP
Catalog No. 501945242
Encompass_Preferred
Description: Stainless Steel Closure. MADE IN THE USASize: 13mmMaterial of Closure: 304 SSTDimensions of Cap: 18/32” OD x 1 1/4” Long, approx.Packed 144 per CaseFingers Grip Tightly after Repeated AutoclavingFor all 13mm OD Culture Tubes and as Flask Closures
Catalog No. 50-194-5242 Supplier Bellco Glass, Inc. Supplier No. 200500013A
Description: Stainless Steel Closure. MADE IN THE USASize: 13mmMaterial of Closure: 304 SSTDimensions of Cap: 18/32” OD x 1 1/4” Long, approx.Packed 144 per CaseFingers Grip Tightly after Repeated AutoclavingFor all 13mm OD Culture Tubes and as Flask Closures