Learn More
Bellco Glass, Inc. Stainless Steel Closure,25mm-12Bg (Lanced)
SDP
Catalog No. 501945249
Encompass_Preferred
STAINLESS STEEL CLOSURES. STAINLESS STEEL CLOSURES CONTINUE TO PROVIDE EXCELLENCE IN CULTURE TUBE AND FLASK CLOSURES -MANUFACTURED FROM TYPE 305 STAINLESS STEEL, THEY CAN BE USED INDEFINITELY WITHOUT CORRODING -THE FINGERS GRIP THE OUTSIDE OF THE TUBE FIRMLY AND WILL NOT LOSE THEIR RESILIENCE THROUGH REPEATED AUTOCLAVING OR DRY HEAT STERILIZATION. MADE IN THE USA CASE Qty: 12.
Catalog No. 50-194-5249 Supplier Bellco Glass, Inc. Supplier No. 200502512
STAINLESS STEEL CLOSURES. STAINLESS STEEL CLOSURES CONTINUE TO PROVIDE EXCELLENCE IN CULTURE TUBE AND FLASK CLOSURES -MANUFACTURED FROM TYPE 305 STAINLESS STEEL, THEY CAN BE USED INDEFINITELY WITHOUT CORRODING -THE FINGERS GRIP THE OUTSIDE OF THE TUBE FIRMLY AND WILL NOT LOSE THEIR RESILIENCE THROUGH REPEATED AUTOCLAVING OR DRY HEAT STERILIZATION. MADE IN THE USA CASE Qty: 12.