Learn More
Bellco Glass, Inc. Stainless Steel Condenser Large Flange - Omni Vessels
SDP
Catalog No. 501944954
Encompass_Preferred
Probe adapters & compression fittings. Bellco compression fittings are designed using our standard 45 mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow pH and do monitoring via insertion of these probes into the sidearms of vessels. Bellco probe adapters are designed to allow introduction of threaded probes into vessels through 200 mm stainless steel headplate or a 45 mm vertical sidearm. Use on the large flange systems - optional accessories. MADE IN THE USA.
Catalog No. 50-194-4954 Supplier Bellco Glass, Inc. Supplier No. 196493006
Probe adapters & compression fittings. Bellco compression fittings are designed using our standard 45 mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow pH and do monitoring via insertion of these probes into the sidearms of vessels. Bellco probe adapters are designed to allow introduction of threaded probes into vessels through 200 mm stainless steel headplate or a 45 mm vertical sidearm. Use on the large flange systems - optional accessories. MADE IN THE USA.