Learn More
Bellco Glass, Inc. Stainless Steel Curve Harvest Tube, 5L 1/4OD x 14"L-Round Bottom Vessels"
SDP
Catalog No. 501944956
Encompass_Preferred
Stainless steel curved harvesting tube for 5l round bottom omni vessels. Manufactured from 316l stainless steel with ra finish of 32 micro inch or better inside and out electropolish finish. MADE IN THE USA. Size: 5l Dimensions: 1/4" x 5/32 x 14 1/4"L
Catalog No. 50-194-4956 Supplier Bellco Glass, Inc. Supplier No. 196495030
Stainless steel curved harvesting tube for 5l round bottom omni vessels. Manufactured from 316l stainless steel with ra finish of 32 micro inch or better inside and out electropolish finish. MADE IN THE USA. Size: 5l Dimensions: 1/4" x 5/32 x 14 1/4"L