Learn More
Bellco Glass, Inc. Stainless Steel Curved Harvest Tube,15L 1/2OD x 22"L - Round Bottom Vessel"
SDP
Catalog No. 501944948
Encompass_Preferred
Stainless steel curved harvesting tube for 15l round bottom omni vessels. Manufactured from 316l stainless steel with ra finish of 32 micro inch or better inside and out electropolish finish. MADE IN THE USA. Size: 15l Dimensions: 1/2" x .430 x 22"
Catalog No. 50-194-4948 Supplier Bellco Glass, Inc. Supplier No. 196491215
Stainless steel curved harvesting tube for 15l round bottom omni vessels. Manufactured from 316l stainless steel with ra finish of 32 micro inch or better inside and out electropolish finish. MADE IN THE USA. Size: 15l Dimensions: 1/2" x .430 x 22"