Learn More
Bellco Glass, Inc. Stainless Steel Curved Harvest Tube,36L 1/2OD x 24" - Round Bottom Vessels"
SDP
Catalog No. 501944949
Encompass_Preferred
Stainless steel curved harvesting tube for 36l round bottom omni vessels. Manufactured from 316l stainless steel with ra finish of 32 micro inch or better inside and out electropolish finish. MADE IN THE USA. Size: 36l Dimensions: 1/2" x .430 x 24"
Catalog No. 50-194-4949 Supplier Bellco Glass, Inc. Supplier No. 196491236
Stainless steel curved harvesting tube for 36l round bottom omni vessels. Manufactured from 316l stainless steel with ra finish of 32 micro inch or better inside and out electropolish finish. MADE IN THE USA. Size: 36l Dimensions: 1/2" x .430 x 24"