Learn More
Bellco Glass, Inc. Stainless Steel Curved Harvest Tube, 5L 1/2OD x 14"L - Round Bottom Vessel"
SDP
Catalog No. 501944945
Encompass_Preferred
Stainless steel curved harvesting tube for 5l round bottom omni vessels. MADE IN THE USA. Manufactured from 316l stainless steel with ra finish of 32 micro inch or better outside only electropolish finish. Size: 5l Dimensions: 1/2" OD x .430"ID x 14 1/4" L
Catalog No. 50-194-4945 Supplier Bellco Glass, Inc. Supplier No. 196491205
Stainless steel curved harvesting tube for 5l round bottom omni vessels. MADE IN THE USA. Manufactured from 316l stainless steel with ra finish of 32 micro inch or better outside only electropolish finish. Size: 5l Dimensions: 1/2" OD x .430"ID x 14 1/4" L