Learn More
Bellco Glass, Inc. Stainless Steel Fitting w/Cap, 1/2in.
SDP
Catalog No. 501945000
Encompass_Preferred
Probe adapters & compression fittings. Bellco compression fittings are designed using our standard 45 mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow pH and do monitoring via insertion of these probes into the sidearms of vessels. Bellco probe adapters are designed to allow introduction of threaded probes into vessels through 200 mm stainless steel headplate or a 45 mm vertical sidearm. MADE IN THE USA. Probe OD: 1/2" Stainless steel fitting w/ cap ½"
Catalog No. 50-194-5000 Supplier Bellco Glass, Inc. Supplier No. 196521070
Probe adapters & compression fittings. Bellco compression fittings are designed using our standard 45 mm blue polypropylene open top caps and white silicone gaskets. A wide variety of fittings are available to allow pH and do monitoring via insertion of these probes into the sidearms of vessels. Bellco probe adapters are designed to allow introduction of threaded probes into vessels through 200 mm stainless steel headplate or a 45 mm vertical sidearm. MADE IN THE USA. Probe OD: 1/2" Stainless steel fitting w/ cap ½"