Learn More
Bellco Glass, Inc. Stainless Steel Harvest Port Assembly , 3/8 For 3L Bioreactor System
SDP
Catalog No. 501944721
Encompass_Preferred
Stainless steel harvest port assembly 3/8 for 3l bioreactor systems. MADE IN THE USA Fits .375" hole in headplate Material: SST 316l ra 32 finish or better Tubing: 316l 1/4" od x 5/32" id Dimension: 10 11/16" oal Insertion length: 9 1/4" Supply with O-ring & SST nut
Catalog No. 50-194-4721 Supplier Bellco Glass, Inc. Supplier No. 196402030
Stainless steel harvest port assembly 3/8 for 3l bioreactor systems. MADE IN THE USA Fits .375" hole in headplate Material: SST 316l ra 32 finish or better Tubing: 316l 1/4" od x 5/32" id Dimension: 10 11/16" oal Insertion length: 9 1/4" Supply with O-ring & SST nut