Learn More
Bellco Glass, Inc. Stainless Steel Hi-Torq Over Head Drive Impeller,8L-FW Full Width
SDP
Catalog No. 501944864
Encompass_Preferred
MADE IN THE USA. Stainless Steel Hi-Torq Over Head Drive Impeller Full Width 8LSpecifications:Hi-Torq SST Over Head Drive Impeller Assembly – Full WidthSize: 8L . For Hi-Torq Motor & Cap Assembly (7774-40115/40230 & 7764-74000)Similar To 1964-60008 But To Accept 1/2″ Shaft From 7764-74000 Cap AssemblyMaterial: 316L SST With Elelctropolish Finish Ra32 Or BetterPaddle Size: 3″ Tall X 8″ Wide
Catalog No. 50-194-4864 Supplier Bellco Glass, Inc. Supplier No. 196474008
MADE IN THE USA. Stainless Steel Hi-Torq Over Head Drive Impeller Full Width 8LSpecifications:Hi-Torq SST Over Head Drive Impeller Assembly – Full WidthSize: 8L . For Hi-Torq Motor & Cap Assembly (7774-40115/40230 & 7764-74000)Similar To 1964-60008 But To Accept 1/2″ Shaft From 7764-74000 Cap AssemblyMaterial: 316L SST With Elelctropolish Finish Ra32 Or BetterPaddle Size: 3″ Tall X 8″ Wide