Learn More
Bellco Glass, Inc. Stainless Steel Hi-Torq Over Head Drive Implr,15L-FW Full Width
SDP
Catalog No. 501944865
Encompass_Preferred
MADE IN THE USA. Hi-torq SST OHD impeller assembly, full widthSize: 15lFor hi-torq motor & cap assembly (7774-40115/40230/ & 7764-74000)Similar to 1964-60015 but to accept 1/2" shaft from 7764-74000 cap assemblyMaterial: 316lPaddle size: 3" tall x 9" wide
Catalog No. 50-194-4865 Supplier Bellco Glass, Inc. Supplier No. 196474015
MADE IN THE USA. Hi-torq SST OHD impeller assembly, full widthSize: 15lFor hi-torq motor & cap assembly (7774-40115/40230/ & 7764-74000)Similar to 1964-60015 but to accept 1/2" shaft from 7764-74000 cap assemblyMaterial: 316lPaddle size: 3" tall x 9" wide