Learn More
Bellco Glass, Inc. Stainless Steel Hi-Torq Over Head Drive Implr,15L-SW Short Width
SDP
Catalog No. 501944868
Encompass_Preferred
MADE IN THE USA. Hi-torq SST OHD impeller assembly, short widthSize: 15lFor hi-torq motor & cap assembly (7774-40115/40230 & 7764-74000)Similar to 1964-60115 but to accept 1/2" shaft from 7764-74000 cap assemblyMaterial: SST 316LPaddle size: 3" tall x 7 1/2" wide
Catalog No. 50-194-4868 Supplier Bellco Glass, Inc. Supplier No. 196474115
Add to Cart
Add to Cart
MADE IN THE USA. Hi-torq SST OHD impeller assembly, short widthSize: 15lFor hi-torq motor & cap assembly (7774-40115/40230 & 7764-74000)Similar to 1964-60115 but to accept 1/2" shaft from 7764-74000 cap assemblyMaterial: SST 316LPaddle size: 3" tall x 7 1/2" wide