Learn More
Bellco Glass, Inc. Stainless Steel Hi-Torq Over Head Drive Implr,36L-FW Full Width
SDP
Catalog No. 501944866
Encompass_Preferred
MADE IN THE USA. Hi-torq SST OHD impeller assembly - full widthSize: 36lFor hi-torq motor & cap assembly (7774-40115/40230 & 7764-74000)Similar to 1964-60036 but to accept 1/2" shaft from 7764-74000 cap assemblyMaterial: 316l SSTPaddle size: 3" tall x 14" wide
Catalog No. 50-194-4866 Supplier Bellco Glass, Inc. Supplier No. 196474036
MADE IN THE USA. Hi-torq SST OHD impeller assembly - full widthSize: 36lFor hi-torq motor & cap assembly (7774-40115/40230 & 7764-74000)Similar to 1964-60036 but to accept 1/2" shaft from 7764-74000 cap assemblyMaterial: 316l SSTPaddle size: 3" tall x 14" wide