Learn More
Bellco Glass, Inc. Stainless Steel Hi-Torq Over Head Drive Implr,36L-SW Short Width
SDP
Catalog No. 501944869
Encompass_Preferred
MADE IN THE USA. Hi-torq SST OHD impeller assembly - short widthSize: 36lFor hi-torq motor & cap assembly (7774-40115/40230 & 7764-74000)Similar to 1964-60136 but to accept 1/2" shaft from 7764-74000 cap assemblyMaterial: 316L SSTPaddle size: 3" tall x 12" wide
Catalog No. 50-194-4869 Supplier Bellco Glass, Inc. Supplier No. 196474136
MADE IN THE USA. Hi-torq SST OHD impeller assembly - short widthSize: 36lFor hi-torq motor & cap assembly (7774-40115/40230 & 7764-74000)Similar to 1964-60136 but to accept 1/2" shaft from 7764-74000 cap assemblyMaterial: 316L SSTPaddle size: 3" tall x 12" wide