Learn More
Bellco Glass, Inc. Stainless Steel Inc Shelf For Bellco Roll-in Inc
SDP
Catalog No. 501946051
Encompass_Preferred
Fits in adjustable clips inside the incubator. Has 1.75" x 0.375" holes for air flow. MADE IN THE USA Material: 304 stainless steel Loads: 120lbs. Supply with 4 shelf brackets Dimensions: 25"x34"
Catalog No. 50-194-6051 Supplier Bellco Glass, Inc. Supplier No. 772840035
Fits in adjustable clips inside the incubator. Has 1.75" x 0.375" holes for air flow. MADE IN THE USA Material: 304 stainless steel Loads: 120lbs. Supply with 4 shelf brackets Dimensions: 25"x34"