Learn More
Bellco Glass, Inc. Stainless Steel Marine Impeller,3OD-LH 3/8" Bore - 1/4-20 set screw
SDP
Catalog No. 501946464
Encompass_Preferred
Impeller with 3"OD and left oriented blades. Single or multiple blade fit onto drive shaft of 3L and 7L motor drive assembly for mammalian cell culture.. MADE IN THE USA
Catalog No. 50-194-6464 Supplier Bellco Glass, Inc. Supplier No. 790910001
Impeller with 3"OD and left oriented blades. Single or multiple blade fit onto drive shaft of 3L and 7L motor drive assembly for mammalian cell culture.. MADE IN THE USA