Learn More
Bellco Glass, Inc. Stainless Steel Marine Impeller,3OD-RH 3/8" Bore - 1/4-20 set screw
SDP
Catalog No. 501946465
Encompass_Preferred
Impeller with 3"OD and right oriented blades. Single or multiple blade fit onto drive shaft of 3L and 7L motor drive assembly for mammalian cell culture.. MADE IN THE USA
Catalog No. 50-194-6465 Supplier Bellco Glass, Inc. Supplier No. 790910002
Impeller with 3"OD and right oriented blades. Single or multiple blade fit onto drive shaft of 3L and 7L motor drive assembly for mammalian cell culture.. MADE IN THE USA