Learn More
Bellco Glass, Inc. Stainless Steel Marine Impeller, 4OD-RH With 1/2"bore & set screw"
SDP
Catalog No. 501946496
Encompass_Preferred
SST Marine Impeller Only. Material: SST 316L with EP finish. 4" OD Prop. Bore size: 1/2". With 1/4-20 sst set screw. Pitch: Right hand. MADE IN THE USA
Catalog No. 50-194-6496 Supplier Bellco Glass, Inc. Supplier No. 790915601
SST Marine Impeller Only. Material: SST 316L with EP finish. 4" OD Prop. Bore size: 1/2". With 1/4-20 sst set screw. Pitch: Right hand. MADE IN THE USA