Learn More
Bellco Glass, Inc. Stainless Steel Marine Shaft Only, 36L With Clip
SDP
Catalog No. 501946204
Encompass_Preferred
Material: 316l stainless steel with electropolish finishDimensions: 1/2" x 16 1/2"Size: 36lIncludes: e-clip for mounting marine prop (1964-73000). MADE IN THE USA
Catalog No. 50-194-6204 Supplier Bellco Glass, Inc. Supplier No. 776476236
Material: 316l stainless steel with electropolish finishDimensions: 1/2" x 16 1/2"Size: 36lIncludes: e-clip for mounting marine prop (1964-73000). MADE IN THE USA