Learn More
Bellco Glass, Inc. Stainless Steel Marine Shaft Only, 5L With Clip
SDP
Catalog No. 501946201
Encompass_Preferred
Manufactured from 316L stainless steel with electropolish finish. Dimensions: 1/2" od x 7 1/2" long Size: 5l Includes: e-clip for mounting marine prop (1964-73000). MADE IN THE USA
Catalog No. 50-194-6201 Supplier Bellco Glass, Inc. Supplier No. 776476205
Manufactured from 316L stainless steel with electropolish finish. Dimensions: 1/2" od x 7 1/2" long Size: 5l Includes: e-clip for mounting marine prop (1964-73000). MADE IN THE USA