Learn More
Bellco Glass, Inc. Stainless Steel Marine Shaft Only, 8L With Clip
SDP
Catalog No. 501946202
Encompass_Preferred
Material: 316l stainless steel with electropolish finishDimensions: 1/2"x 7 1/2"Size: 8lIncludes: e-clip for mounting marine prop (1964-73000). MADE IN THE USA
Catalog No. 50-194-6202 Supplier Bellco Glass, Inc. Supplier No. 776476208
Material: 316l stainless steel with electropolish finishDimensions: 1/2"x 7 1/2"Size: 8lIncludes: e-clip for mounting marine prop (1964-73000). MADE IN THE USA