Learn More
Bellco Glass, Inc. Stainless Steel Metal Rack,35mm For Auto Blot Tubes
SDP
Catalog No. 501946507
Encompass_Preferred
SST Bottle Rack, 3x4. Dimensions: 9 1/2" x 6 3/4" x 4 1/2"Holds 12 bottles. For 35mm ID Bellco Auto-Blot Tubes:7910-35300, 7910-35225, 7910-35150, 7910-35010. With Two SST Handles. MADE IN THE USA
Catalog No. 50-194-6507 Supplier Bellco Glass, Inc. Supplier No. 791003040
Add to Cart
Add to Cart
SST Bottle Rack, 3x4. Dimensions: 9 1/2" x 6 3/4" x 4 1/2"Holds 12 bottles. For 35mm ID Bellco Auto-Blot Tubes:7910-35300, 7910-35225, 7910-35150, 7910-35010. With Two SST Handles. MADE IN THE USA