Learn More
Bellco Glass, Inc. Stainless Steel Over Head Drive Impeller Assembly , 15L-FW Full Width
SDP
Catalog No. 501944833
Encompass_Preferred
Manufactured from 316l stainless steel for standard bioreactor vessels. Use with over head drive motors. MADE IN THE USA. PADDLE L X W (IN): 3 X 9.
Catalog No. 50-194-4833 Supplier Bellco Glass, Inc. Supplier No. 196460015
Manufactured from 316l stainless steel for standard bioreactor vessels. Use with over head drive motors. MADE IN THE USA. PADDLE L X W (IN): 3 X 9.