Learn More
Bellco Glass, Inc. Stainless Steel Over Head Drive Impeller Assembly , 6L Full Width
SDP
Catalog No. 501944831
Encompass_Preferred
Manufactured from 316l stainless steel for standard bioreactor vessels. Use with over head drive motors. MADE IN THE USA. PADDLE L X W (IN): 3 X 6 3/4".
Catalog No. 50-194-4831 Supplier Bellco Glass, Inc. Supplier No. 196460006
Manufactured from 316l stainless steel for standard bioreactor vessels. Use with over head drive motors. MADE IN THE USA. PADDLE L X W (IN): 3 X 6 3/4".