Learn More
Bellco Glass, Inc. Stainless Steel Over Head Drive Short Width Impeller Assembly , 15L Short Wdth
SDP
Catalog No. 501944837
Encompass_Preferred
Short width stainless steel folding impeller assembly for standard bioreactor vessels 15l. MADE IN THE USA. 316 l stainless steel Ra finish 32 micro inch or better (wxl) 3" x 7 1/2"
Catalog No. 50-194-4837 Supplier Bellco Glass, Inc. Supplier No. 196460115
Short width stainless steel folding impeller assembly for standard bioreactor vessels 15l. MADE IN THE USA. 316 l stainless steel Ra finish 32 micro inch or better (wxl) 3" x 7 1/2"