Learn More
Bellco Glass, Inc. Stainless Steel Over Head Drive Short Width Impeller Assembly , 8L Short Width
SDP
Catalog No. 501944836
Encompass_Preferred
Short width stainless steel folding impeller assembly for standard bioreactor vessels 8l. MADE IN THE USA. 316 l stainless steel Ra finish 32 micro inch or better (wxl) 3" x 6"
Catalog No. 50-194-4836 Supplier Bellco Glass, Inc. Supplier No. 196460108
Short width stainless steel folding impeller assembly for standard bioreactor vessels 8l. MADE IN THE USA. 316 l stainless steel Ra finish 32 micro inch or better (wxl) 3" x 6"