Learn More
Bellco Glass, Inc. Stainless Steel Pipet Can,5x5X16 cm w/ Silicone Pads
SDP
Catalog No. 501944601
Encompass_Preferred
Heavy gauge rectangular constructionfor easy stacking and storage. Thesepipette cans can be sterilized byautoclave or dry heat. (do not exceed400 degrees F). Includes silicone padon the inside of top and bottom toprotect pipette tips.--Satin finish surface of entire can forconvenient marking or coding.--Silicone pads on inside of top and bottomprotect pipette tips.--Material: 304 stainless steel .031” thickMADE IN THE USA
Catalog No. 50-194-4601 Supplier Bellco Glass, Inc. Supplier No. 121711106
Heavy gauge rectangular constructionfor easy stacking and storage. Thesepipette cans can be sterilized byautoclave or dry heat. (do not exceed400 degrees F). Includes silicone padon the inside of top and bottom toprotect pipette tips.--Satin finish surface of entire can forconvenient marking or coding.--Silicone pads on inside of top and bottomprotect pipette tips.--Material: 304 stainless steel .031” thickMADE IN THE USA