Learn More
Bellco Glass, Inc. Stainless Steel Pipet Can,6.6x6.6x46 cm w/out Silicone Pads
SDP
Catalog No. 501944608
Encompass_Preferred
Heavy gauge rectangular constructionfor easy stacking and storage. Thesepipette cans can be sterilized byautoclave or dry heat. (do not exceed400 degrees F). Includes silicone padon the inside of top and bottom toprotect pipette tips.--Satin finish surface of entire can forconvenient marking or coding.--W/O Silicone pads --Material: 304 stainless steel .031” thickMADE IN THE USA
Catalog No. 50-194-4608 Supplier Bellco Glass, Inc. Supplier No. 121790218
Heavy gauge rectangular constructionfor easy stacking and storage. Thesepipette cans can be sterilized byautoclave or dry heat. (do not exceed400 degrees F). Includes silicone padon the inside of top and bottom toprotect pipette tips.--Satin finish surface of entire can forconvenient marking or coding.--W/O Silicone pads --Material: 304 stainless steel .031” thickMADE IN THE USA