Learn More
Bellco Glass, Inc. Stainless Steel Pipet Can,6.6x6.6x46cm w/ Silicone Pads
SDP
Catalog No. 501944605
Encompass_Preferred
Heavy gauge rectangular constructionfor easy stacking and storage. Thesepipette cans can be sterilized byautoclave or dry heat. (do not exceed400 degrees F). Includes silicone padon the inside of top and bottom toprotect pipette tips.--Satin finish surface of entire can forconvenient marking or coding.--Silicone pads on inside of top and bottomprotect pipette tips.--Material: 304 stainless steel .031” thickMADE IN THE USA
Catalog No. 50-194-4605 Supplier Bellco Glass, Inc. Supplier No. 121722218
Heavy gauge rectangular constructionfor easy stacking and storage. Thesepipette cans can be sterilized byautoclave or dry heat. (do not exceed400 degrees F). Includes silicone padon the inside of top and bottom toprotect pipette tips.--Satin finish surface of entire can forconvenient marking or coding.--Silicone pads on inside of top and bottomprotect pipette tips.--Material: 304 stainless steel .031” thickMADE IN THE USA