Learn More
Bellco Glass, Inc. Stainless Steel Pitched Blade Impeller, 3L For 3L Bioreactor Systems
SDP
Catalog No. 501944732
Encompass_Preferred
SST Pitched Blade Impeller MADE IN THE USA Material: 316l SST with electropolish finishEnglish threadOD: 1 3/4"For 5/16" od shaftsUsually requires 2 each
Catalog No. 50-194-4732 Supplier Bellco Glass, Inc. Supplier No. 196402200
SST Pitched Blade Impeller MADE IN THE USA Material: 316l SST with electropolish finishEnglish threadOD: 1 3/4"For 5/16" od shaftsUsually requires 2 each