Learn More
Bellco Glass, Inc. Stainless Steel Platform Tray,10x10" "
SDP
Catalog No. 501946108
Encompass_Preferred
For use with sticky matt tape (7740-40000). Dimensions: 10"x10", .050" thick w/ 1/2" lip with pegs. Will fit any Bellco rocker or shaker. Plain tray - no rubber matt.. MADE IN THE USA
Catalog No. 50-194-6108 Supplier Bellco Glass, Inc. Supplier No. 774002525
For use with sticky matt tape (7740-40000). Dimensions: 10"x10", .050" thick w/ 1/2" lip with pegs. Will fit any Bellco rocker or shaker. Plain tray - no rubber matt.. MADE IN THE USA