Learn More
Bellco Glass, Inc. Stainless Steel Platform Tray, 12x12" "
SDP
Catalog No. 501946110
Encompass_Preferred
For use with sticky matt tape (7740-40000). Dimensions: 12"x12", .050" thick w/ 1/2" lip with pegs. Will fit any Bellco rocker or shaker. Plain tray - no rubber matt.. MADE IN THE USA
Catalog No. 50-194-6110 Supplier Bellco Glass, Inc. Supplier No. 774003030
For use with sticky matt tape (7740-40000). Dimensions: 12"x12", .050" thick w/ 1/2" lip with pegs. Will fit any Bellco rocker or shaker. Plain tray - no rubber matt.. MADE IN THE USA