Learn More
Bellco Glass, Inc. Stainless Steel Removable Baffle For Large Flange Vessels
SDP
Catalog No. 501944806
Encompass_Preferred
Removable baffle for large flange vessels. MADE IN THE USA. Material: SST 36l with electropolish finish Dimensions: 1 1/2" wide x 10 1/2" long Fits on any 1/4" od SST tube inside vessel Recommend: 2 per vessel Z-shaped
Catalog No. 50-194-4806 Supplier Bellco Glass, Inc. Supplier No. 196453400
Removable baffle for large flange vessels. MADE IN THE USA. Material: SST 36l with electropolish finish Dimensions: 1 1/2" wide x 10 1/2" long Fits on any 1/4" od SST tube inside vessel Recommend: 2 per vessel Z-shaped