Learn More
Bellco Glass, Inc. Stainless Steel Sample Port Assembly , 3/8 For 7L Bioreactor Systems"
SDP
Catalog No. 501944729
Encompass_Preferred
SST Sample Port Assyembly for 7l bioreactor systems - English threads MADE IN THE USAMaterial: 316l SST with electropolish finish - inside & outTubing specifications: .250" od x .152" idOverall length: 14 5/16"Insertion length: 12 7/8"Supply with SST 3/8-24 nut & o-ring
Catalog No. 50-194-4729 Supplier Bellco Glass, Inc. Supplier No. 196402090
SST Sample Port Assyembly for 7l bioreactor systems - English threads MADE IN THE USAMaterial: 316l SST with electropolish finish - inside & outTubing specifications: .250" od x .152" idOverall length: 14 5/16"Insertion length: 12 7/8"Supply with SST 3/8-24 nut & o-ring