Learn More
Bellco Glass, Inc. Stainless Steel Sample Port Assembly , 3/8 For 3L Bioreactor System
SDP
Catalog No. 501944720
Encompass_Preferred
Ideal for 3l bioreactors. Fits 3/8" hole Material: stainless steel 316l Dimensions: 9 3/16" oal Insertion length: 7 3/4" Tube: 1/4" od x 5/32" id with ra finish With hose barb Supply with: o-ring & stainless steel nut
Catalog No. 50-194-4720 Supplier Bellco Glass, Inc. Supplier No. 196402020
Ideal for 3l bioreactors. Fits 3/8" hole Material: stainless steel 316l Dimensions: 9 3/16" oal Insertion length: 7 3/4" Tube: 1/4" od x 5/32" id with ra finish With hose barb Supply with: o-ring & stainless steel nut