Learn More
Bellco Glass, Inc. Stainless Steel Shaft Assembly ,100mL
SDP
Catalog No. 501945154
Encompass_Preferred
Description: 100mL Stainless Steel Replacement Assembly. MADE IN THE USAThis assembly consists of; SST shaft, glass impeller shaft bearing, silicone tubing & compression fitting.Specifications of SST Shaft:Material: 316L SST with Electroplish FinishRa Finish: 32 micro inch or betterDimensions: 6.35mm (1/4”) OD x 120.6mm (4 ¾”) longSpecifications of Glass Bearing:Material: Type 1 Class A Borosilicate GlassDimensions: 9mm OD x 12mm long turn down to 6.35mm endSpecifications of Silicone Tubing:Material: Clear Silicone Tubing, FDA approveDimensions: 4.9mm OD x 2.6mm ID x 15.9mm long to fit shaft & bearingSpecifications Compression Fitting:Material: White PolypropyleneDimensions: 6.35mm ID Compression fitting to accept 6.35mm (1/4”) OD SST Shaft
Catalog No. 50-194-5154 Supplier Bellco Glass, Inc. Supplier No. 196960100
Description: 100mL Stainless Steel Replacement Assembly. MADE IN THE USAThis assembly consists of; SST shaft, glass impeller shaft bearing, silicone tubing & compression fitting.Specifications of SST Shaft:Material: 316L SST with Electroplish FinishRa Finish: 32 micro inch or betterDimensions: 6.35mm (1/4”) OD x 120.6mm (4 ¾”) longSpecifications of Glass Bearing:Material: Type 1 Class A Borosilicate GlassDimensions: 9mm OD x 12mm long turn down to 6.35mm endSpecifications of Silicone Tubing:Material: Clear Silicone Tubing, FDA approveDimensions: 4.9mm OD x 2.6mm ID x 15.9mm long to fit shaft & bearingSpecifications Compression Fitting:Material: White PolypropyleneDimensions: 6.35mm ID Compression fitting to accept 6.35mm (1/4”) OD SST Shaft