Learn More
Bellco Glass, Inc. Stainless Steel Shaft Assembly , 15-36L
SDP
Catalog No. 501945158
Encompass_Preferred
Stainless Steel Shaft Assembly The Bellco Stainless Steel Shaft Conversion Kit is designed to convert your standard glass shaft to stainless steel on 1967 and1965 Spinner Flasks. Sizes range from 100 mL to 36 liters for 1965 spinners and 100 to 3000 mL for 1967 spinner flasks. Material: 316L Stainless. MADE IN THE USASteel and polypropylene compression fitting. Size: 15-36l
Catalog No. 50-194-5158 Supplier Bellco Glass, Inc. Supplier No. 196961500
Stainless Steel Shaft Assembly The Bellco Stainless Steel Shaft Conversion Kit is designed to convert your standard glass shaft to stainless steel on 1967 and1965 Spinner Flasks. Sizes range from 100 mL to 36 liters for 1965 spinners and 100 to 3000 mL for 1967 spinner flasks. Material: 316L Stainless. MADE IN THE USASteel and polypropylene compression fitting. Size: 15-36l