Learn More
Bellco Glass, Inc. Stainless Steel Sparge Assembly w/Frit, 3/8 For 7L Bioreactor System
SDP
Catalog No. 501944723
Encompass_Preferred
316l stainless steel sparger assembly with stainless steel frit for 7 liter bioreactor system. MADE IN THE USA English threadsMaterial: 316l SST with electropolish finish ra 32 or betterTubing specifications: .250" od x .152" id Overall length: 10 43/64" Inseration Length: 9 1/4" With SST Frit - 10µ Supply with SST Nut & O-ring
Catalog No. 50-194-4723 Supplier Bellco Glass, Inc. Supplier No. 196402045
316l stainless steel sparger assembly with stainless steel frit for 7 liter bioreactor system. MADE IN THE USA English threadsMaterial: 316l SST with electropolish finish ra 32 or betterTubing specifications: .250" od x .152" id Overall length: 10 43/64" Inseration Length: 9 1/4" With SST Frit - 10µ Supply with SST Nut & O-ring