Learn More
Bellco Glass, Inc. Thermowell Assemblies are used to hold inside one tube the RTD temperature sensor that monitors the temperature within the vessel in conjunction with a temperature controller. There are also tubes for addition of media and vent tube. Fit through a 5/1
SDP
Supplier:  Bellco Glass, Inc. 780311530
Encompass_Preferred
Thermowell Assemblies are used to hold inside one tube the RTD temperature sensor that monitors the temperature within the vessel in conjunction with a temperature controller. There are also tubes for addition of media and vent tube. Fit through a 5/1
Catalog No. 50-194-6277