Learn More
Bellco Glass, Inc. Stainless Steel Thermowell, 5L
SDP
Catalog No. 501946280
Encompass_Preferred
Thermowell Assemblies are used to hold inside one tube the RTD temperature sensor that monitors the temperature within the vessel in conjunction with a temperature controller. There are also tubes for addition of media and vent tube. Fit through a A 5/16" FITTING ON THE STAINLESS STEEL HEADPLATE. . MADE IN THE USA
Catalog No. 50-194-6280 Supplier Bellco Glass, Inc. Supplier No. 780311533
Thermowell Assemblies are used to hold inside one tube the RTD temperature sensor that monitors the temperature within the vessel in conjunction with a temperature controller. There are also tubes for addition of media and vent tube. Fit through a A 5/16" FITTING ON THE STAINLESS STEEL HEADPLATE. . MADE IN THE USA