Learn More
Bellco Glass, Inc. Thermowell Assemblies are used to hold inside one tube the RTD temperature sensor that monitors the temperature within the vessel in conjunction with a temperature controller. There are also tubes for addition of media and vent tube. Fit through a 5/1
SDP
Catalog No. 501946279
Encompass_Preferred
Thermowell Assemblies are used to hold inside one tube the RTD temperature sensor that monitors the temperature within the vessel in conjunction with a temperature controller. There are also tubes for addition of media and vent tube. Fit through a 5/1
Catalog No. 50-194-6279 Supplier Bellco Glass, Inc. Supplier No. 780311532
Thermowell Assemblies are used to hold inside one tube the RTD temperature sensor that monitors the temperature within the vessel in conjunction with a temperature controller. There are also tubes for addition of media and vent tube. Fit through a 5/1