Learn More
Bellco Glass, Inc. Stainless Steel Tube Rack, 13mm-40P 4x10 Spacing
SDP
Catalog No. 501945300
Encompass_Preferred
Will not rust or deteriorate in water baths or co2 atmospheres. Stainless steel material Each opening rack 9/16" (14.3mm) Tube size: 13mm Spacing: 4x10 holds 40 tubes Dimensions: 6 1/2" x 2 3/4" x 2" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .135" OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each. MADE IN THE USA
Catalog No. 50-194-5300 Supplier Bellco Glass, Inc. Supplier No. 202713040
Add to Cart
Add to Cart
Will not rust or deteriorate in water baths or co2 atmospheres. Stainless steel material Each opening rack 9/16" (14.3mm) Tube size: 13mm Spacing: 4x10 holds 40 tubes Dimensions: 6 1/2" x 2 3/4" x 2" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .135" OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each. MADE IN THE USA