Learn More
Bellco Glass, Inc. Stainless Steel Tube Rack,13mm-72P 6x12 Spacing
SDP
Catalog No. 501945301
Encompass_Preferred
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 19/32" (15mm) Tube size: 13mm Spacing: 6x12 holds 72 tubes Dimensions: 8 1/6" x 4 1/8" x 2" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .135"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each
Catalog No. 50-194-5301 Supplier Bellco Glass, Inc. Supplier No. 202713072
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 19/32" (15mm) Tube size: 13mm Spacing: 6x12 holds 72 tubes Dimensions: 8 1/6" x 4 1/8" x 2" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .135"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each