Learn More
Bellco Glass, Inc. Stainless Steel Tube Rack,13mm-90P 6x15 Spacing
SDP
Catalog No. 501945302
Encompass_Preferred
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 9/16" (14.3mm) Tube size: 13mm Spacing: 6x15 holds 90 tubes Dimensions: 9 5/8" x 4" x 2 1/4" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .135"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each
Catalog No. 50-194-5302 Supplier Bellco Glass, Inc. Supplier No. 202713090
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 9/16" (14.3mm) Tube size: 13mm Spacing: 6x15 holds 90 tubes Dimensions: 9 5/8" x 4" x 2 1/4" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .135"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each