Learn More
Bellco Glass, Inc. Stainless Steel Tube Rack,16mm-120P 10x12 Spacing
SDP
Catalog No. 501945308
Encompass_Preferred
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 11/16" (17.5mm) Tube size: 16mm Spacing: 10x12 holds 120 tubes Dimensions: 9 1/4" x 7 5/8" x 3" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .135"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each
Catalog No. 50-194-5308 Supplier Bellco Glass, Inc. Supplier No. 202716120
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 11/16" (17.5mm) Tube size: 16mm Spacing: 10x12 holds 120 tubes Dimensions: 9 1/4" x 7 5/8" x 3" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .135"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each