Learn More
Bellco Glass, Inc. Stainless Steel Tube Rack,16mm-150P 6x25 Spacing
SDP
Catalog No. 501945309
Encompass_Preferred
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 11/16" (17.5mm) Tube size: 16mm Spacing: 6x25 holds 150 tubes Dimensions: 19" x 4 3/4" x 3" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .135"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each
Catalog No. 50-194-5309 Supplier Bellco Glass, Inc. Supplier No. 202716150
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 11/16" (17.5mm) Tube size: 16mm Spacing: 6x25 holds 150 tubes Dimensions: 19" x 4 3/4" x 3" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .135"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each