Learn More
Bellco Glass, Inc. Stainless Steel Tube Rack,20-22mm-12P 3x4 Spacing
SDP
Catalog No. 501945313
Encompass_Preferred
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 15/16" (23.8mm) Tube size: 20-22mm Spacing: 3x4 holds 12 tubes Dimensions: 4 5/16" X 3 1/4" X 3 15/16" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .135"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each
Catalog No. 50-194-5313 Supplier Bellco Glass, Inc. Supplier No. 202720012
Add to Cart
Add to Cart
Will not rust or deteriorate in water baths or co2 atmospheres. Manufactured from 303 stainless steel with electropolish finish. MADE IN THE USA. Each opening rack 15/16" (23.8mm) Tube size: 20-22mm Spacing: 3x4 holds 12 tubes Dimensions: 4 5/16" X 3 1/4" X 3 15/16" WIRE SPECIFICATIONS: SIDE & UPRIGHTS: .135"OD WIRE, CROSS WIRE: 0.063" OD WIRE, BTTM MESH: 1/4" Minimum order requirement: 5 each